Gene Transcripts

Gene Symbol
C5orf42
Entrez Gene ID
65250
Gene Name
chromosome 5 open reading frame 42
Gene Aliases
Hug, JBTS17, OFD6
Chromosome Location
Chr. 5: 37063928 - 37249428 on Build GRCh38
UniGene ID
Hs.586199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023073.3 NP_075561.3 39 8031
XM_005248345.3 XP_005248402.1 39 8019
XM_005248346.3 XP_005248403.1 39 7936
XM_005248347.3 XP_005248404.1 39 7936
XM_005248349.3 XP_005248406.1 39 7936
XM_005248350.3 XP_005248407.1 39 7808
XM_006714491.3 XP_006714554.1 13 2378
XM_011514085.2 XP_011512387.1 39 7938
XM_011514086.2 XP_011512388.1 39 7935
XM_011514087.2 XP_011512389.1 39 8024
XM_011514088.2 XP_011512390.1 39 8024
XM_011514089.2 XP_011512391.1 39 8024
XM_011514090.2 XP_011512392.1 39 7802
XM_011514092.2 XP_011512394.1 39 8024
XM_011514094.2 XP_011512396.1 24 5073
XM_017009760.1 XP_016865249.1 38 7750
XM_017009761.1 XP_016865250.1 38 7718
XM_017009762.1 XP_016865251.1 35 7477
XM_017009763.1 XP_016865252.1 35 7350
XM_017009764.1 XP_016865253.1 35 7311
XM_017009765.1 XP_016865254.1 39 7719
XM_017009766.1 XP_016865255.1 25 5201
GenBank mRNA AK096581.1 - 1 120
AL833065.2 - 14 3262
BC144069.1 - 24 4944 9052589
BC144070.1 - 24 4813
BC150594.1 - 24 4944 9056964
BX647602.1 - 6 659
BX649121.1 - 1 42

View Full Product Details