Gene Transcripts

Gene Symbol
C4orf36
Entrez Gene ID
132989
Gene Name
chromosome 4 open reading frame 36
Gene Aliases
Chromosome Location
Chr. 4: 86876205 - 86897256 on Build GRCh38
UniGene ID
Hs.339646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144645.3 NP_653246.1 3, 4 538
XM_011531610.2 XP_011529912.1 3, 4 425
XM_011531611.2 XP_011529913.1 3, 4 647
XM_011531612.2 XP_011529914.1 3, 4 357
XM_011531613.2 XP_011529915.1 3, 4 892
XM_011531615.2 XP_011529917.1 3, 4 377
XM_017007746.1 XP_016863235.1 3, 4 319
GenBank mRNA BC016746.1 - 3, 4 514
BX090513.1 - 2, 3 279

View Full Product Details