Gene Transcripts

Gene Symbol
C11orf88
Entrez Gene ID
399949
Gene Name
chromosome 11 open reading frame 88
Gene Aliases
Chromosome Location
Chr. 11: 111514785 - 111537031 on Build GRCh38
UniGene ID
Hs.411600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100388.1 NP_001093858.1 6 655
NM_207430.2 NP_997313.2 7 736
GenBank mRNA AK128145.1 - 6 3041
BC039505.2 - 6 640

View Full Product Details