Gene Transcripts

Gene Symbol
CAMK4
Entrez Gene ID
814
Gene Name
calcium/calmodulin dependent protein kinase IV
Gene Aliases
CaMK IV, CaMK-GR, caMK
Chromosome Location
Chr. 5: 111223653 - 111498503 on Build GRCh38
UniGene ID
Hs.438801
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323374.1 NP_001310303.1 12 1802
NM_001323375.1 NP_001310304.1 12 1834
NM_001323376.1 NP_001310305.1 9 1804
NM_001323377.1 NP_001310306.1 10 1562
NM_001744.5 NP_001735.1 11 1950
GenBank mRNA AK311662.1 - 9 1659
AL833368.1 - 2 1712
BC025687.1 - 11 1792 5205478
BX374853.2 - 1 704

View Full Product Details