Gene Transcripts

Gene Symbol
C17orf75
Entrez Gene ID
64149
Gene Name
chromosome 17 open reading frame 75
Gene Aliases
NJMU-R1, SRI2
Chromosome Location
Chr. 17: 32328441 - 32350047 on Build GRCh38
UniGene ID
Hs.655257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022344.3 NP_071739.2 5 575
XM_005258022.3 XP_005258079.1 6 593
XM_017024940.1 XP_016880429.1 5 444
XM_017024941.1 XP_016880430.1 5 442
GenBank mRNA AF052093.1 - 5 502
AF305686.1 - 5 531
BC026017.2 - 5 521 3352761
BC035715.1 - 5 529 5588164
BC047325.1 - 5 515 5493477
BC054035.1 - 5 509 6143106
BP308900.1 - 5 560

View Full Product Details