Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317958.1 | NP_001304887.1 | 11 | 1597 | |
NM_001317959.1 | NP_001304888.1 | 13 | 1983 | ||
NM_152572.2 | NP_689785.1 | 11 | 1590 | ||
XM_005272169.2 | XP_005272226.1 | 11 | 1129 | ||
XM_006716965.2 | XP_006717028.1 | 9 | 902 | ||
XM_011518278.2 | XP_011516580.1 | 10 | 970 | ||
XM_017014308.1 | XP_016869797.1 | 5 | 652 | ||
GenBank mRNA | AK057266.1 | - | 11 | 1063 | |
AK093446.1 | - | 11 | 1105 | ||
AK292186.1 | - | 11 | 1075 | ||
BC034776.1 | - | 11 | 1087 | 5175757 | |
BC050576.1 | - | 11 | 1590 | 5744517 |