Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016429.3 | NP_057513.1 | 2 | 178 | |
XM_011524874.2 | XP_011523176.1 | 2 | 177 | ||
GenBank mRNA | AB037938.1 | - | 2 | 162 | |
AB047849.1 | - | 2 | 160 | ||
BC015924.1 | - | 2 | 154 | 4850757 | |
HY179834.1 | - | 2 | 173 |