Gene Transcripts
- Gene Symbol
- CYP3A7
- Entrez Gene ID
- 1551
- Gene Name
- cytochrome P450 family 3 subfamily A member 7
- Gene Aliases
- CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA
- Chromosome Location
- Chr. 7: 99705037 - 99735200 on Build GRCh38
- UniGene ID
- Hs.111944
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000765.4 |
NP_000756.3
|
13 |
1757 |
|
GenBank mRNA |
D00408.1 |
- |
13 |
1653 |
|
View Full Product Details