Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109878.1 | NP_001103348.1 | 3 | 434 | |
NM_001109879.1 | NP_001103349.1 | 3 | 434 | ||
NM_001303475.1 | NP_001290404.1 | 1 | 125 | ||
NM_016954.2 | NP_058650.1 | 2 | 327 | ||
XM_005262136.2 | XP_005262193.1 | 3 | 462 | ||
XM_006724657.3 | XP_006724720.1 | 3 | 463 | ||
XM_011530972.1 | XP_011529274.1 | 2 | 1004 | ||
GenBank mRNA | AF251684.1 | - | 1 | 116 | |
AK097514.1 | - | 3 | 434 | ||
AY035371.1 | - | 2 | 306 | ||
BC014194.2 | - | 2 | 327 | 3952809 | |
DB456367.1 | - | 3 | 428 |