Gene Transcripts
- Gene Symbol
- CACFD1
- Entrez Gene ID
- 11094
- Gene Name
- calcium channel flower domain containing 1
- Gene Aliases
- C9orf7, D9S2135, FLOWER
- Chromosome Location
- Chr. 9: 133459965 - 133470787 on Build GRCh38
- UniGene ID
- Hs.62003
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135775.2 |
NP_001129247.1
|
3, 4 |
413 |
|
NM_017586.3 |
NP_060056.1
|
4, 5 |
539 |
|
XM_011518185.2 |
XP_011516487.1
|
4, 5 |
2096 |
|
XM_011518186.1 |
XP_011516488.1
|
4, 5 |
592 |
|
GenBank mRNA |
AJ011373.1 |
- |
4, 5 |
495 |
|
AK074852.1 |
- |
4, 5 |
517 |
|
AK075416.1 |
- |
3, 4 |
411 |
|
AK075548.1 |
- |
4, 5 |
507 |
|
AK304524.1 |
- |
5, 6 |
497 |
|
AK311325.1 |
- |
4, 5 |
595 |
|
AY358452.1 |
- |
4, 5 |
487 |
|
BC030558.1 |
- |
4, 5 |
505 |
5223979 |
DA699497.1 |
- |
3, 4 |
350 |
|
DB165724.1 |
- |
4, 5 |
539 |
|
View Full Product Details