Gene Transcripts

Gene Symbol
Asb2
Entrez Gene ID
299266
Gene Name
ankyrin repeat and SOCS box-containing 2
Gene Aliases
Chromosome Location
Chr. 6: 127212325 - 127248451 on Build Rnor_6.0
UniGene ID
Rn.98677
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011984.1 NP_001011984.1 7, 8 1227
XM_006240464.2 XP_006240526.1 5, 6 1872
GenBank mRNA BC085882.1 - 7, 8 1227

View Full Product Details