Gene Transcripts

Gene Symbol
Alg13
Entrez Gene ID
300284
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
Chromosome Location
Chr. X: 115561358 - 115569256 on Build Rnor_6.0
UniGene ID
Rn.29819
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013951.1 NP_001013973.1 4 1555
GenBank mRNA BC088233.1 - 4 1555
FQ216283.1 - 4 1566
FQ229089.1 - 4 1581

View Full Product Details