Gene Transcripts

Gene Symbol
AADACL4
Entrez Gene ID
343066
Gene Name
arylacetamide deacetylase like 4
Gene Aliases
Chromosome Location
Chr. 1: 12644547 - 12667086 on Build GRCh38
UniGene ID
Hs.209954
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013630.1 NP_001013652.1 4 1355
XM_017001152.1 XP_016856641.1 4 1280
XM_017001153.1 XP_016856642.1 2 1317

View Full Product Details