Gene Transcripts

Gene Symbol
Ankrd54
Entrez Gene ID
362957
Gene Name
ankyrin repeat domain 54
Gene Aliases
RGD1309552
Chromosome Location
Chr. 7: 120282423 - 120295646 on Build Rnor_6.0
UniGene ID
Rn.28758
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025285.1 NP_001020456.1 7 875
XM_006242012.2 XP_006242074.1 7 948
GenBank mRNA BC093616.1 - 7 875

View Full Product Details