Gene Transcripts

Gene Symbol
OR5H14
Entrez Gene ID
403273
Gene Name
olfactory receptor family 5 subfamily H member 14
Gene Aliases
Chromosome Location
Chr. 3: 98149386 - 98150318 on Build GRCh38
UniGene ID
Hs.554714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005514.1 NP_001005514.1 1 637
GenBank mRNA BC136984.1 - 1 697

View Full Product Details