Gene Transcripts

Gene Symbol
Vom1r41
Entrez Gene ID
494261
Gene Name
vomeronasal 1 receptor 41
Gene Aliases
V1rg5
Chromosome Location
Chr. 1: 67742949 - 67743932 on Build Rnor_6.0
UniGene ID
Rn.129425
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008909.1 NP_001008909.1 1 477
GenBank mRNA AY510299.1 - 1 477

View Full Product Details