Gene Transcripts

Gene Symbol
Zfand3
Entrez Gene ID
361816
Gene Name
zinc finger, AN1-type domain 3
Gene Aliases
Tex27
Chromosome Location
Chr. 20: 8694004 - 8892570 on Build Rnor_6.0
UniGene ID
Rn.136939
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012175.1 NP_001012175.1 2, 3 230
XM_008772822.1 XP_008771044.1 2, 3 124
GenBank mRNA BC085956.1 - 2, 3 230

View Full Product Details