Gene Transcripts

Gene Symbol
Phf7
Entrez Gene ID
364510
Gene Name
PHD finger protein 7
Gene Aliases
Chromosome Location
Chr. 16: 7323527 - 7336230 on Build Rnor_6.0
UniGene ID
Rn.24375
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012211.1 NP_001012211.1 11 1197
XM_006252632.2 XP_006252694.1 9 1015
GenBank mRNA BC079288.1 - 11 1197

View Full Product Details