Gene Transcripts

Gene Symbol
Nol12
Entrez Gene ID
362955
Gene Name
nucleolar protein 12
Gene Aliases
Nop25, RGD1305327
Chromosome Location
Chr. 7: 120161238 - 120166852 on Build Rnor_6.0
UniGene ID
Rn.17454
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012747.2 NP_001012765.1 1 92
GenBank mRNA AY917132.1 - 1 70
BC103643.1 - 1 92 7301307
FQ227943.1 - 1 140

View Full Product Details