Gene Transcripts

Gene Symbol
Reep6
Entrez Gene ID
362835
Gene Name
receptor accessory protein 6
Gene Aliases
Dp1l1
Chromosome Location
Chr. 7: 12240059 - 12246999 on Build Rnor_6.0
UniGene ID
Rn.9353
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013218.1 NP_001013236.1 6 1913
GenBank mRNA BC083830.1 - 6 1913

View Full Product Details