Gene Transcripts

Gene Symbol
Mageh1
Entrez Gene ID
367767
Gene Name
melanoma antigen, family H, 1
Gene Aliases
Chromosome Location
Chr. X: 19243759 - 19245015 on Build Rnor_6.0
UniGene ID
Rn.56595
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013250.1 NP_001013268.1 1 856
GenBank mRNA BC087574.1 - 1 856

View Full Product Details