Gene Transcripts

Gene Symbol
Bag4
Entrez Gene ID
361167
Gene Name
BCL2-associated athanogene 4
Gene Aliases
Chromosome Location
Chr. 16: 71057709 - 71075874 on Build Rnor_6.0
UniGene ID
Rn.163329
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025130.1 NP_001020301.1 4 1637
GenBank mRNA BC083608.1 - 4 1637

View Full Product Details