Gene Transcripts

Gene Symbol
Slfnl1
Entrez Gene ID
500540
Gene Name
schlafen-like 1
Gene Aliases
Chromosome Location
Chr. 5: 139466895 - 139473599 on Build Rnor_6.0
UniGene ID
Rn.47152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024347.1 NP_001019518.1 3 1312
XM_008764060.1 XP_008762282.1 2 1621
XM_008764061.1 XP_008762283.1 4 2846
XM_008764062.1 XP_008762284.1 4 2846
XM_008764063.1 XP_008762285.1 2 1621
GenBank mRNA BC079280.1 - 3 1312

View Full Product Details