Gene Transcripts

Gene Symbol
Apof
Entrez Gene ID
500761
Gene Name
apolipoprotein F
Gene Aliases
Chromosome Location
Chr. 7: 2689501 - 2690938 on Build Rnor_6.0
UniGene ID
Rn.101696
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024351.1 NP_001019522.1 2 979
GenBank mRNA AY325229.1 - 728
BC088170.1 - 2 979 7322324

View Full Product Details