Gene Transcripts

Gene Symbol
Mfsd3
Entrez Gene ID
500899
Gene Name
major facilitator superfamily domain containing 3
Gene Aliases
Chromosome Location
Chr. 7: 117763399 - 117765904 on Build Rnor_6.0
UniGene ID
Rn.86351
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024908.1 NP_001020079.1 5 1483
XM_006241859.2 XP_006241921.1 5 1972
GenBank mRNA BC097424.1 - 5 1483

View Full Product Details