Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027599.3 | NP_081875.1 | 2 | 2433 | |
GenBank mRNA | AK029548.1 | - | 2 | 2435 | |
AV255931.2 | - | 1 | 655 | ||
BC139095.1 | - | 2 | 2058 | 8862117 | |
BC139096.1 | - | 2 | 2058 | 8862118 |