Gene Transcripts

Gene Symbol
Plekhf1
Entrez Gene ID
308543
Gene Name
pleckstrin homology domain containing, family F (with FYVE domain) member 1
Gene Aliases
Chromosome Location
Chr. 1: 94609041 - 94616572 on Build Rnor_6.0
UniGene ID
Rn.9380
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013148.1 NP_001013166.1 1 861
XM_006228905.2 XP_006228967.1 2 956
GenBank mRNA BC079354.1 - 1 861

View Full Product Details