Gene Transcripts

Gene Symbol
Adal
Entrez Gene ID
311352
Gene Name
adenosine deaminase-like
Gene Aliases
Chromosome Location
Chr. 3: 113091782 - 113109841 on Build Rnor_6.0
UniGene ID
Rn.204498
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014047.1 NP_001014069.1 5 803
XM_008762166.1 XP_008760388.1 10 1281
XM_008762167.1 XP_008760389.1 8 811
XM_008762168.1 XP_008760390.1 6 712
XM_008762169.1 XP_008760391.1 9 1017
GenBank mRNA BC089959.1 - 5 803

View Full Product Details