Gene Transcripts

Gene Symbol
Stap1
Entrez Gene ID
305269
Gene Name
signal transducing adaptor family member 1
Gene Aliases
RGD1311584, Stap-1
Chromosome Location
Chr. 14: 23576084 - 23604983 on Build Rnor_6.0
UniGene ID
Rn.205047
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025115.1 NP_001020286.1 1, 2 158
XM_006250812.1 XP_006250874.1 1, 2 307
GenBank mRNA BC091249.1 - 1, 2 158

View Full Product Details