Gene Transcripts

Gene Symbol
RBM22
Entrez Gene ID
55696
Gene Name
RNA binding motif protein 22
Gene Aliases
Cwc2, ZC3H16, fSAP47
Chromosome Location
Chr. 5: 150690790 - 150701107 on Build GRCh38
UniGene ID
Hs.713564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018047.2 NP_060517.1 4 279
GenBank mRNA AK001152.1 - 4 234
AL136933.1 - 4 208
BC003402.1 - 4 212 3449258
DA534416.1 - 4 280

View Full Product Details