Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
Chromosome Location
Chr. 2: 189674292 - 189763059 on Build GRCh38
UniGene ID
Hs.60772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144708.3 NP_653309.3 21 4028
XM_011510673.2 XP_011508975.1 21 4547
XM_011510674.1 XP_011508976.1 21 4514
XM_011510675.2 XP_011508977.1 20 4442
XM_011510676.2 XP_011508978.1 21 4547
XM_011510677.2 XP_011508979.1 21 4547
XM_011510679.2 XP_011508981.1 19 4118
XM_011510685.2 XP_011508987.1 21 4103
XM_011510686.2 XP_011508988.1 17 4040
XM_011510687.2 XP_011508989.1 20 3911
XM_011510691.2 XP_011508993.1 17 2733
XM_011510692.2 XP_011508994.1 19 3747
XM_011510693.2 XP_011508995.1 18 3643
XM_011510694.2 XP_011508996.1 17 3462
XM_017003415.1 XP_016858904.1 16 3242
XM_017003416.1 XP_016858905.1 18 3654
XM_017003418.1 XP_016858907.1 16 3357
XM_017003419.1 XP_016858908.1 16 3324
XM_017003420.1 XP_016858909.1 16 3260
GenBank mRNA AJ549812.1 - 20 3727
AK058144.1 - 9 1380
BC044907.1 - 17 3521 4836761
BC047413.1 - 24 3592

View Full Product Details