Gene Transcripts

Gene Symbol
NUDT11
Entrez Gene ID
55190
Gene Name
nudix hydrolase 11
Gene Aliases
APS1, ASP1, DIPP3b, DIPP3beta, hDIPP3beta
Chromosome Location
Chr. X: 51490011 - 51496607 on Build GRCh38
UniGene ID
Hs.200016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018159.3 NP_060629.2 2 1140
GenBank mRNA AK001490.1 - 2 1027
BC009942.2 - 2 1094 4303422

View Full Product Details