Gene Transcripts

Gene Symbol
SEC61A2
Entrez Gene ID
55176
Gene Name
Sec61 translocon alpha 2 subunit
Gene Aliases
Chromosome Location
Chr. 10: 12129641 - 12169958 on Build GRCh38
UniGene ID
Hs.112955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142627.2 NP_001136099.1 4, 5 375
NM_001142628.1 NP_001136100.1 3, 4 309
NM_018144.3 NP_060614.2 4, 5 375
NR_024576.2 4, 5 375
NR_024577.2 5, 6 508
GenBank mRNA AF346603.1 - 4, 5 316
AK022640.1 - 4, 5 310
AK057532.1 - 5, 6 449
AK292295.1 - 4, 5 325
AK301841.1 - 3, 4 262
BC026179.1 - 4, 5 325 4823130
BP212860.1 - 4, 5 375

View Full Product Details