Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042635.1 | NP_001036100.1 | 7 | 506 | |
NM_015514.1 | NP_056329.1 | 7 | 506 | ||
GenBank mRNA | AK022215.1 | - | 4 | 1071 | |
AK124318.1 | - | 3 | 2107 | ||
AK291875.1 | - | 7 | 499 | ||
AL050003.1 | - | 7 | 494 | ||
BC030817.1 | - | 7 | 492 | 4768089 |