Gene Transcripts

Gene Symbol
TMEM184C
Entrez Gene ID
55751
Gene Name
transmembrane protein 184C
Gene Aliases
TMEM34
Chromosome Location
Chr. 4: 147617371 - 147637273 on Build GRCh38
UniGene ID
Hs.203896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018241.2 NP_060711.2 1, 2 700
GenBank mRNA AK001708.1 - 1, 2 643
BC046128.1 - 1, 2 702 5750613

View Full Product Details