Gene Transcripts

Gene Symbol
TEX36
Entrez Gene ID
387718
Gene Name
testis expressed 36
Gene Aliases
C10orf122, bA383C5.1
Chromosome Location
Chr. 10: 125576522 - 125683144 on Build GRCh38
UniGene ID
Hs.148259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318133.1 NP_001305062.1 4 554
GenBank mRNA BC127766.1 - 4 435
BC127767.1 - 4 439 40127345
BX336391.2 - 4 516

View Full Product Details