Gene Transcripts

Gene Symbol
C11orf96
Entrez Gene ID
387763
Gene Name
chromosome 11 open reading frame 96
Gene Aliases
AG2
Chromosome Location
Chr. 11: 43942556 - 43943883 on Build GRCh38
UniGene ID
Hs.530443
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145033.1 NP_001138505.1 1 1291
GenBank mRNA BC052560.1 - 1 1257

View Full Product Details