Gene Transcripts

Gene Symbol
C2orf80
Entrez Gene ID
389073
Gene Name
chromosome 2 open reading frame 80
Gene Aliases
GONDA1
Chromosome Location
Chr. 2: 208165347 - 208190258 on Build GRCh38
UniGene ID
Hs.198416
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099334.2 NP_001092804.1 2, 3 216
XM_017004074.1 XP_016859563.1 2, 3 229
XM_017004076.1 XP_016859565.1 2, 3 204
GenBank mRNA BC035737.1 - 2, 3 216

View Full Product Details