Gene Transcripts

Gene Symbol
ANKDD1B
Entrez Gene ID
728780
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
Chromosome Location
Chr. 5: 75611289 - 75671846 on Build GRCh38
UniGene ID
Hs.370455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011543618.2 XP_011541920.1 1 86
XM_011543623.2 XP_011541925.1 1 86
XM_017009814.1 XP_016865303.1 1 86
XM_017009815.1 XP_016865304.1 1 86
XM_017009816.1 XP_016865305.1 1 86
XM_017009817.1 XP_016865306.1 1 86

View Full Product Details