Gene Transcripts

Gene Symbol
AKR1B15
Entrez Gene ID
441282
Gene Name
aldo-keto reductase family 1 member B15
Gene Aliases
AKR1B10L, AKR1B10L, AK1R1B7, AKR1R1B7
Chromosome Location
Chr. 7: 134549097 - 134579841 on Build GRCh38
UniGene ID
Hs.729418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080538.2 NP_001074007.2 3 415
XM_011516241.2 XP_011514543.2 1 40
XM_017012224.1 XP_016867713.1 1 168

View Full Product Details