Gene Transcripts

Gene Symbol
ATXN7L3
Entrez Gene ID
56970
Gene Name
ataxin 7 like 3
Gene Aliases
Chromosome Location
Chr. 17: 44191805 - 44199976 on Build GRCh38
UniGene ID
Hs.512651
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098833.1 NP_001092303.1 12 1352
NM_020218.1 NP_064603.1 12 1373
XM_011525038.2 XP_011523340.1 12 2024
XM_011525044.2 XP_011523346.1 12 1967
XM_011525046.2 XP_011523348.1 12 1946
XM_017024880.1 XP_016880369.1 13 1602
XM_017024881.1 XP_016880370.1 13 1901
XM_017024882.1 XP_016880371.1 12 2003
XM_017024883.1 XP_016880372.1 13 1605
XM_017024884.1 XP_016880373.1 12 1627
XM_017024885.1 XP_016880374.1 13 1548
XM_017024886.1 XP_016880375.1 13 1527
GenBank mRNA AK056002.1 - 9 896
AL390158.1 - 7 782
BC024695.2 - 5 710
BC037418.1 - 12 1280

View Full Product Details