Gene Transcripts

Gene Symbol
PRR12
Entrez Gene ID
57479
Gene Name
proline rich 12
Gene Aliases
KIAA1205
Chromosome Location
Chr. 19: 49591177 - 49626439 on Build GRCh38
UniGene ID
Hs.590971
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017027031.1 XP_016882520.1 6 4643
GenBank mRNA AB033031.1 - 5 2375
BC034003.1 - 3 741

View Full Product Details