Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
319089
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
C14orf25, NCRNA00291
Chromosome Location
Chr. 14: 37595199 - 37842628 on Build GRCh38
UniGene ID
Hs.659936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310135.1 NP_001297064.1 2, 3 898
XM_011537432.2 XP_011535734.1 2, 3 898
XM_017021254.1 XP_016876743.1 2, 3 898
XM_017021255.1 XP_016876744.1 2, 3 898
XM_017021257.1 XP_016876746.1 2, 3 898

View Full Product Details