Gene Transcripts

Gene Symbol
RIPPLY3
Entrez Gene ID
53820
Gene Name
ripply transcriptional repressor 3
Gene Aliases
DSCR6
Chromosome Location
Chr. 21: 37006150 - 37019659 on Build GRCh38
UniGene ID
Hs.254560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317768.1 NP_001304697.1 4 780
NM_001317777.1 NP_001304706.1 3 660
NM_018962.2 NP_061835.1 4 1010
GenBank mRNA AB037158.1 - 4 1010
AB037159.1 - 4 780
AB037160.1 - 3 660
AB037161.1 - 1 646

View Full Product Details