Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001006933.1 | NP_001006934.1 | 3 | 1066 | |
NM_032926.2 | NP_116315.1 | 3 | 1054 | ||
GenBank mRNA | AF163263.1 | - | 3 | 1003 | |
AK098494.1 | - | 3 | 997 | ||
BC007622.1 | - | 3 | 1045 | 3355622 | |
BC008703.1 | - | 3 | 1054 | 2966921 |