Gene Transcripts

Gene Symbol
Vom1r84
Entrez Gene ID
494248
Gene Name
vomeronasal 1 receptor 84
Gene Aliases
V1rc43
Chromosome Location
Chr. 4: 88206954 - 88208035 on Build Rnor_6.0
UniGene ID
Rn.129656
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009528.1 NP_001009528.1 1 782
XM_008762962.1 XP_008761184.1 2 855
GenBank mRNA AY510312.1 - 1 782

View Full Product Details