Gene Transcripts

Gene Symbol
Rbm22
Entrez Gene ID
307410
Gene Name
RNA binding motif protein 22
Gene Aliases
Chromosome Location
Chr. 18: 55828158 - 55838966 on Build Rnor_6.0
UniGene ID
Rn.105130
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025676.1 NP_001020847.1 4 279
GenBank mRNA BC097991.1 - 4 279

View Full Product Details