Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031637.1 | NP_001026807.1 | 18 | 2915 | |
XM_006245927.2 | XP_006245989.1 | 19 | 4023 | ||
XM_006245928.2 | XP_006245990.1 | 19 | 4007 | ||
XM_008767554.1 | XP_008765776.1 | 19 | 4637 | ||
XM_008767555.1 | XP_008765777.1 | 18 | 3031 | ||
XM_008767556.1 | XP_008765778.1 | 18 | 2896 | ||
XM_008767557.1 | XP_008765779.1 | 18 | 3031 | ||
GenBank mRNA | AB196699.1 | - | 18 | 2915 |