Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142391.2 | NP_001135863.1 | 4 | 719 | |
NM_001142392.2 | NP_001135864.1 | 3 | 806 | ||
NM_019848.4 | NP_062822.1 | 2 | 1041 | ||
XM_005277913.4 | XP_005277970.1 | 2 | 1463 | ||
XM_006724847.3 | XP_006724910.1 | 2 | 984 | ||
XM_006724848.3 | XP_006724911.1 | 3 | 897 | ||
XM_011531200.2 | XP_011529502.2 | 2 | 1016 | ||
XM_011531201.2 | XP_011529503.1 | 2 | 720 | ||
GenBank mRNA | BC004966.1 | - | 4 | 643 | |
BX401493.2 | - | 3 | 778 |