Gene Transcripts

Gene Symbol
Atad3a
Entrez Gene ID
298682
Gene Name
ATPase family, AAA domain containing 3A
Gene Aliases
Atad3
Chromosome Location
Chr. 5: 173189590 - 173209768 on Build Rnor_6.0
UniGene ID
Rn.17450
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034922.1 NP_001030094.1 15, 16 1666
GenBank mRNA BC105762.1 - 15, 16 1666

View Full Product Details